Skip to Content
Merck
All Photos(2)

Key Documents

8.40085

Sigma-Aldrich

(2-Ethoxyethyl) methacrylate

(stabilized) for synthesis

Synonym(s):

(2-Ethoxyethyl) methacrylate, Methacrylic acid 2-ethoxyethyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
₹4,868.36

₹4,868.36


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 ML
₹4,868.36

About This Item

Empirical Formula (Hill Notation):
C8H14O3
CAS Number:
Molecular Weight:
158.19
MDL number:
UNSPSC Code:
12352108
EC Index Number:
219-135-3
NACRES:
NA.22

₹4,868.36


Please contact Customer Service for Availability

Request a Bulk Order

vapor pressure

<1 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

209 °C

potency

>2000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

bp

185 °C

transition temp

flash point 69.5 °C

solubility

17.05 g/L

density

0.96 g/cm3 at 20 °C

storage temp.

2-8°C

SMILES string

O(CCOC(=O)C(=C)C)CC

InChI

1S/C8H14O3/c1-4-10-5-6-11-8(9)7(2)3/h2,4-6H2,1,3H3

InChI key

SFPNZPQIIAJXGL-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.963 - 0.967
Identity (IR): passes test
The material is stabilised with MEHQ. At time of analytical testing a content of >10 ppm stabiliser was determined.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service