Skip to Content
Merck
All Photos(1)

Documents

8.18888

Sigma-Aldrich

Xenon difluoride

for synthesis

Synonym(s):

Xenon difluoride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
F2Xe
CAS Number:
Molecular Weight:
169.29
MDL number:
UNSPSC Code:
12352307
EC Index Number:
237-251-2
NACRES:
NA.22

vapor pressure

3.8 hPa ( 25 °C)

Quality Level

form

crystals

mp

130-135 °C (sublimed)

storage temp.

2-8°C

InChI

1S/F2Xe/c1-3-2

InChI key

IGELFKKMDLGCJO-UHFFFAOYSA-N

Application


  • Organic Laboratory Applications: Discusses how xenon difluoride interacts with organic substrates, highlighting its reaction mechanisms and versatility in organic synthesis (Ramsden, 2014).

  • Aromatic Compounds Fluorination: Investigates the fluorination of aromatic compounds using xenon difluoride, emphasizing its advantages over elemental fluorine for safer and more controlled reactions (Fedorov et al., 2015).

  • Microfabrication: Describes the use of xenon difluoride for dry etching in microfabrication, particularly for creating microelectromechanical systems by etching sacrificial silicon layers (Rowlands, 2015).

  • Excitation Studies: Explores the excitation of Europium(III) in the presence of xenon difluoride, showing how xenon difluoride can assist in producing electronically excited states (Masyagutova et al., 2024).

  • Etching of Sacrificial Layers: Details the application of xenon difluoride in the etching of sacrificial layers for the fabrication of microelectromechanical devices, demonstrating its effectiveness in creating complex structures (Vahanissi, 2019).

Analysis Note

Assay (iodometric): ≥ 98.0 %

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Ox. Sol. 2 - Skin Corr. 1B

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service