Skip to Content
Merck
All Photos(1)

Key Documents

8.06908

Sigma-Aldrich

2-Anilinoethanol

for synthesis

Synonym(s):

2-Anilinoethanol, N-(2-Hydroxyethyl)aniline, N-Phenylethanolamine, 2-(Phenylamino)ethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H11NO
CAS Number:
Molecular Weight:
137.18
MDL number:
UNSPSC Code:
12352104
EC Index Number:
204-588-1

vapor pressure

1.46 hPa ( 100 °C)

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

410 °C (DIN 51794)

potency

2230 mg/kg LD50, oral (Rat)
68 mg/kg LD50, skin (Rabbit)

expl. lim.

1-6.8 % (v/v)

pH

10.2 (20 °C, 10 g/L in H2O)

mp

-30 °C

transition temp

flash point 153 °C (DIN 51758)

solubility

4.5 g/L

density

1.08 g/cm3 at 20 °C

InChI

1S/C8H11NO/c10-7-6-9-8-4-2-1-3-5-8/h1-5,9-10H,6-7H2

InChI key

MWGATWIBSKHFMR-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.094 - 1.096
Identity (IR): passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Aquatic Chronic 3 - Eye Dam. 1 - STOT RE 2

Target Organs

Blood

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(C)

113 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service