Skip to Content
Merck
All Photos(1)

Key Documents

6.18686

Supelco

Manganese (II) Chloride tetrahydrate

PFD

Synonym(s):

Manganese(II) chloride tetrahydrate

Sign Into View Organizational & Contract Pricing

Select a Size

IN26186860500
₹876.00

₹876.00


Estimated to ship on31 March 2025Details


Request a Bulk Order

Select a Size

Change View
IN26186860500
₹876.00

About This Item

Empirical Formula (Hill Notation):
Cl2Mn · 4H2O
CAS Number:
Molecular Weight:
197.91
MDL number:
UNSPSC Code:
12352302
grade:
for analytical purposes

₹876.00


Estimated to ship on31 March 2025Details


Request a Bulk Order

grade

for analytical purposes

SMILES string

[Mn+2].[Cl-].[Cl-].O.O.O.O

InChI

1S/2ClH.Mn.4H2O/h2*1H;;4*1H2/q;;+2;;;;/p-2

InChI key

CNFDGXZLMLFIJV-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service