Skip to Content
Merck
All Photos(1)

Key Documents

6.18569

Supelco

Octane-1-sulphonic Acid Sodium Salt Monohydrate

Synonym(s):

Octane-1-sulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

IN26185690025
₹2,180.00

₹2,180.00


Estimated to ship on22 March 2025Details


Request a Bulk Order

Select a Size

Change View
IN26185690025
₹2,180.00

About This Item

Empirical Formula (Hill Notation):
C8H17NaO3S
CAS Number:
Molecular Weight:
216.27
MDL number:
UNSPSC Code:
12161902
grade:
for analytical purposes

₹2,180.00


Estimated to ship on22 March 2025Details


Request a Bulk Order

grade

for analytical purposes

SMILES string

[Na+].[S](=O)(=O)([O-])CCCCCCCC

InChI

1S/C8H18O3S.Na/c1-2-3-4-5-6-7-8-12(9,10)11;/h2-8H2,1H3,(H,9,10,11);/q;+1/p-1

InChI key

HRQDCDQDOPSGBR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service