Skip to Content
Merck
All Photos(1)

Key Documents

6.18300

Sigma-Aldrich

Acetonitrile

for chromatography (gradient grade 2.5)

Synonym(s):

Acetonitrile, ACN, Cyanomethane, Ethyl nitrile, Methyl cyanide

Sign Into View Organizational & Contract Pricing

Select a Size

IN26183002500
₹2,479.99

₹2,479.99


Estimated to ship on15 February 2025Details


Request a Bulk Order

Select a Size

Change View
IN26183002500
₹2,479.99

About This Item

Linear Formula:
CH3CN
CAS Number:
Molecular Weight:
41.05
Beilstein:
741857
MDL number:
UNSPSC Code:
12191502
grade:
for chromatography (gradient grade 2.5)
Assay:
≥99.9% (GC)
technique(s):
HPLC: suitable
application(s):
general analytical
bp:
81-82 °C (lit.)
vapor pressure:
72.8 mmHg ( 20 °C)

₹2,479.99


Estimated to ship on15 February 2025Details


Request a Bulk Order

grade

for chromatography (gradient grade 2.5)

Quality Level

vapor density

1.41 (vs air)

vapor pressure

72.8 mmHg ( 20 °C)

Assay

≥99.9% (GC)

form

liquid

autoignition temp.

973 °F

expl. lim.

16 %

technique(s)

HPLC: suitable

refractive index

n20/D 1.344 (lit.)

bp

81-82 °C (lit.)

mp

−45 °C (lit.)

solubility

water: soluble (1,000 g/l at 25 °C (77 °F))

density

0.786 g/mL at 25 °C (lit.)

application(s)

general analytical

SMILES string

CC#N

InChI

1S/C2H3N/c1-2-3/h1H3

InChI key

WEVYAHXRMPXWCK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acetonitrile is commonly used as a key component in the mobile phase for gradient elution chromatography. It is used in various chromatographic techniques, such as high-performance liquid chromatography (HPLC) and liquid chromatography-mass spectrometry (LC-MS).[1]

Application

Acetonitrile can be used as a mobile phase:
  • Buffer in the liquid chromatographic method for determination of polyamines, amino acids, metanephrines, and catecholamine compounds in human body fluids.
  • Component to facilitate the fast gradient elution separations of polar neutral active pharmaceutical ingredients (APIs).[2]

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development and validation of a liquid chromatography-tandem mass spectrometry (LC-MS/MS) procedure for screening of urine specimens for 100 analytes relevant in drug-facilitated crime (DFC)
Remane D, et al.
Analytical and Bioanalytical Chemistry, 406, 4411-4424 (2014)
Fast gradient screening of pharmaceuticals with 5 cm long, narrow bore reversed-phase columns packed with sub-3 ?m core?shell and sub-2 ?m totally porous particles
Fekete S, et al.
Talanta, 84(2), 416-423 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service