Skip to Content
Merck
All Photos(1)

Key Documents

6.18099

Supelco

Formic acid

purified grade, ~90%

Synonym(s):

Formic acid

Sign Into View Organizational & Contract Pricing

Select a Size

IN26180990500
₹747.99

₹747.99


Estimated to ship on26 March 2025Details


Request a Bulk Order

Select a Size

Change View
IN26180990500
₹747.99

About This Item

Linear Formula:
HCOOH
CAS Number:
Molecular Weight:
46.03
Beilstein:
1209246
MDL number:
UNSPSC Code:
12352106
grade:
purified grade

₹747.99


Estimated to ship on26 March 2025Details


Request a Bulk Order

grade

purified grade

vapor density

1.6 (vs air)

vapor pressure

44.8 mmHg ( 20 °C)

Assay

~90%

autoignition temp.

1004 °F

expl. lim.

57 %

refractive index

n20/D 1.370 (lit.)

bp

100-101 °C (lit.)

mp

8.2-8.4 °C (lit.)

density

1.22 g/mL at 25 °C (lit.)

SMILES string

OC=O

InChI

1S/CH2O2/c2-1-3/h1H,(H,2,3)

InChI key

BDAGIHXWWSANSR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

121.1 °F

Flash Point(C)

49.5 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service