Skip to Content
Merck
All Photos(1)

Key Documents

6.17961

Supelco

Molecular sieve 4A x 1.5 mm

Sign Into View Organizational & Contract Pricing

Select a Size

IN26179610250
₹1,170.00

₹1,170.00


In StockDetails


Request a Bulk Order

Select a Size

Change View
IN26179610250
₹1,170.00

About This Item

UNSPSC Code:
23201100

₹1,170.00


In StockDetails


Request a Bulk Order

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service