613571
TIRAP Inhibitor Peptide, Control, Cell-Permeable
The TIRAP Inhibitor Peptide, Control, Cell-Permeable serves as a control for TIRAP Inhibitor Peptide. This small molecule/inhibitor is primarily used for Inflammation/Immunology applications.
Synonym(s):
TIRAP Inhibitor Peptide, Control, Cell-Permeable, Mal Peptide, MyD88-Adapter Like Peptide, Toll-interleukin 1 Receptor (TIR) domain-containing Adapter Protein Peptide, Ant-Tirap 151-138, TIRAP Peptide, Control, (RQIKIWFQNRRMKWKKSVIAGGPAADRLQL), Mal Peptide, MyD88-Adapter Like Peptide, Toll-interleukin 1 Receptor (TIR) domain-containing Adapter Protein Peptide, Ant-Tirap151-138, TIRAP Peptide, Control, (RQIKIWFQNRRMKWKKSVIAGGPAADRLQL)
About This Item
Recommended Products
Quality Level
Assay
≥97% (HPLC)
form
lyophilized solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
desiccated (hygroscopic)
protect from light
color
white
solubility
DMSO: 5 mg/mL
shipped in
wet ice
storage temp.
−20°C
General description
Biochem/physiol Actions
Control for TIRAP inhibitor
Packaging
Warning
Sequence
Physical form
Reconstitution
Other Notes
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service