Skip to Content
Merck
All Photos(1)

Key Documents

6.09973

Supelco

Hydrochloric acid

c(HCl) = 0.1mol/l (0.1N), concentrate for preparing 0.5 L volumetric solution for titration, Titriplex®

Synonym(s):

Hydrochloric acid solution, Hydrogen chloride solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HCl
CAS Number:
Molecular Weight:
36.46
MDL number:
UNSPSC Code:
12352301

Product Name

Hydrochloric acid, for 500 ml, c(HCl) = 0.1mol/l (0.1N) Titrisol

SMILES string

Cl

InChI

1S/ClH/h1H

InChI key

VEXZGXHMUGYJMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Unlocking high-performance HCl adsorption at elevated temperatures: This research demonstrates the use of hydrochloric acid solution for synthesizing robust Ca-Mg-Al mixed oxides. These materials are crucial in environmental applications for adsorbing acidic gases, showcasing hydrochloric acid′s role in developing solutions to combat air pollution (Cao et al., 2024).

  • Sustainable and Green Corrosion Inhibition of Mild Steel: Hydrochloric acid is utilized here to test new, environmentally friendly corrosion inhibitors. This is vital in industries where maintaining material integrity against chemical attack is crucial, highlighting hydrochloric acid′s role in innovative material science applications (Rizi et al., 2023).

  • Synthesis of Zinc Oxide Nanorods from Zinc Borate Precursor: This study uses hydrochloric acid in the synthesis of zinc oxide nanorods, important for supercapacitor applications. It underscores the critical role of hydrochloric acid in nanomaterials research, pushing the boundaries of energy storage technology (Lefdhil et al., 2023).

  • An improved digestion and analysis procedure for silicon in plant tissue: Hydrochloric acid is employed to digest plant tissues for silicon content analysis, enhancing agricultural and biological research methodologies. This application reflects the importance of hydrochloric acid in scientific studies that require precise chemical processing (Langenfeld and Bugbee, 2023).

  • N-N(+) Bond-Forming Intramolecular Cyclization of O-Tosyloxy β-Aminopropioamidoximes and Ion Exchange Reaction: In this research, hydrochloric acid plays a key role in organic synthesis, demonstrating its versatility in pharmaceutical compound development. The study provides insights into the synthesis of complex organic compounds, which are vital in medicinal chemistry (Kayukova et al., 2023).

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service