559404
SB 239063
A potent, cell-permeable, reversible, and ATP-competitive inhibitor of the α- and β-isoforms (IC₅₀ = 44 nM), but not the γ- or δ-isoform (IC₅₀ > 100 µM), of MAP kinase p38.
Synonym(s):
SB 239063, trans-1-(4-Hydroxycyclohexyl)-4-(4-fluorophenyl)-5-(2-methoxypyrimidin-4-yl)imidazole, p38 MAP Kinase Inhibitor XV
About This Item
Recommended Products
Quality Level
Assay
≥97% (HPLC)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
color
white
solubility
DMSO: 0.5-1 mg/mL
shipped in
ambient
storage temp.
−20°C
InChI
1S/C20H21FN4O2/c1-27-20-22-11-10-17(24-20)19-18(13-2-4-14(21)5-3-13)23-12-25(19)15-6-8-16(26)9-7-15/h2-5,10-12,15-16,26H,6-9H2,1H3
InChI key
ZQUSFAUAYSEREK-UHFFFAOYSA-N
General description
Biochem/physiol Actions
MAPKp38α & β
Packaging
Warning
Preparation Note
Reconstitution
Other Notes
Ju, H., et al. 2002. J. Pharmacol. Exp. Ther.301, 15.
Legos, J.J., et al. 2002. Eur. J. Pharmacol.447, 37.
Barone, F.C., et al. 2001. J. Pharmacol. Exp. Ther.296, 312.
Ward, K.W., et al. 2001. Pharm. Res.18, 1336.
Underwood, D.C., et al. 2000. J. Pharmacol. Exp. Ther.293, 281.
Underwood, D.C., et al. 2000. Am. J. Physiol. Lung Cell Mol. Physiol.279, L895.
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service