Skip to Content
Merck
All Photos(1)

Documents

526559

Sigma-Aldrich

PI3Kδ Inhibitor, SW30

The PI3Kδ Inhibitor, SW30 controls the biological activity of PI3Kδ. This small molecule/inhibitor is primarily used for Membrane applications.

Synonym(s):

PI3Kδ Inhibitor, SW30, 2-((4-amino-3-(3-hydroxyprop-1-ynyl)-1H-pyrazolo[3,4-d]pyrimidin-1-yl)methyl)-5-methyl-3-o-tolylquinazolin-4(3H)-one, PI 3-K Inhibitor XIV

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H21N7O2
Molecular Weight:
451.48
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

>95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A cell permeable, ATP-competitive and selective tolyl quinazolinone inhibitor of PI 3-Kδ (IC50 = 7 nM, 1300 nM, 85000 nM, and 740 nM for PI 3-Kδ PI 3-Kγ PI 3-Kα and PI 3-Kβ, respectively) in an enzymatic assay with an EC50 = 16 nM for PI 3-Kδ and 2.7 µM for PI 3-Kγ in a cell based assay. It elicits an inflammatory marker suppression effect in co-cultures of HUVEC and PBMCs under superantigen stimulated conditions. This compound, at concentrations between 370 nM and 10 µM, leads to a dose-dependent decrease in E-selectin expression. Furthermore, it is not cytotoxic to HUVEC cells.
A cell permeable, ATP-competitive and selective tolyl quinazolinone inhibitor of PI 3-Kδ (IC50 = 7 nM, 1300 nM, 85000 nM, and 740 nM for PI 3-Kδ, PI 3-Kγ PI 3-Kα and PI 3-Kβ, respectively) in an enzymatic assay with an EC50 = 16 nM for PI 3-Kδ and 2.7 µM for PI 3-Kγ in a cell based assay. It elicits an inflammatory marker suppression effect in co-cultures of HUVEC and PBMCs under superantigen stimulated conditions. This compound, at concentrations between 370 nM and 10 µM, leads to a dose-dependent decrease in E-selectin expression. Furthermore, it is not cytotoxic to HUVEC cells.

Warning

Toxicity: Regulatory Review (Z)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Williams, O., et al. 2010. Chem Biol.17, 123.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wuguang Ji et al.
Experimental and therapeutic medicine, 23(6), 414-414 (2022-05-24)
Atherosclerosis is a cardiovascular disease, which is characterized by the interaction between carbohydrates, lipids, cells and various other molecules and genetic factors. Previous studies have demonstrated that resveratrol (RV) served protective roles in numerous types of human disease by regulating

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service