Skip to Content
Merck
All Photos(1)

Key Documents

5.08053

Sigma-Aldrich

Mcl-1 Inhibitor II

Synonym(s):

Mcl-1 Inhibitor II, 7-((4-Ethylpiperazin-1-yl)(4-(trifluoromethyl)phenyl)methyl)quinolin-8-ol, Myeloid Cell Leukemia-1 Inhibitor II, Myeloid Cell Leukemia-1 Inhibitor II, 7-((4-Ethylpiperazin-1-yl)(4-(trifluoromethyl)phenyl)methyl)quinolin-8-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H24F3N3O
CAS Number:
Molecular Weight:
415.45
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥95% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 25 mg/mL

storage temp.

2-8°C

SMILES string

FC(F)(F)c1ccc(cc1)C(N4CCN(CC4)CC)c2c(c3ncccc3cc2)O

InChI key

NGAPBLRRJSKIRT-UHFFFAOYSA-N

General description

A cell-permeable hydroxynaphthalenylthioacetate that targets Mcl-1 BH3-binding groove and selectively competes against 8 nM FITC-AHA-BIM142-161 for Mcl-1, but not Bcl-xL, binding (IC50 = 310 nM vs >40 µM; [Mcl-1] & [Bcl-xL] = 6.25 nM), displaying little inhibitory potency toward CYP450 isoforms 1A2/2D6/3A4/2C9 (IC50 = >10 µM). Antiproliferation potencies against 2 murine lymphoma cell lines (GI50 = 300 nM/ MCl1-1780 & 1.1 µM/Bcl2-1863) and 11 human cancer lines of various origins (GI50 from 1.6 to 25 µM) correlate well with intracellular BH3 domain peptide-binding capacity. Likewise, greater apoptosis induction is observed in MCl1-1780 than the highly Bcl-2/Bcl-xL primed Bcl2-1863 cultures upon drug treatment. Stability studies using murine and human liver microsome preparations reveal t1/2 values of 40 min and 55 min, respectively.
A cell-permeable hydroxynaphthalenylthioacetate that targets Mcl-1 BH3-binding groove and selectively competes against 8 nM FITC-AHA-BIM142-161 for Mcl-1, but not Bcl-xL, binding (IC50 = 310 nM vs >40 µM; [Mcl-1] & [Bcl-xL] = 6.25 nM). Antiproliferation potencies against 2 murine lymphoma cell lines and 11 human cancer lines of various origins (GI50 from 0.3 to 25 µM) correlate well with intracellular BH3 domain peptide-binding capacity. Likewise, greater apoptosis induction is observed in MCl1-1780 than the highly Bcl-2/Bcl-xL primed Bcl2-1863 cultures upon drug treatment.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Mcl-1
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Richard, D.J., et al. 2013. Bioorg. Med. Chem.21, 6642.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service