Skip to Content
Merck
All Photos(1)

Documents

5.05224

Sigma-Aldrich

Necrostatin-1

InSolution, ≥95%, 25 mM in DMSO, required for the biosynthesis of sialic acid-containing complex carbohydrates

Synonym(s):

InSolution Necrostatin-1, 5-((1H-Indol-3-yl)methyl)-3-methyl-2-thioxoimidazolidin-4-one; 5-(Indol-3-ylmethyl)-(2-thio-3-methyl)hydantoin, MTH-Trp, Methyl-thiohydantoin-tryptophan, Nec-1, Nec1, Necrosome Inhibitor I, RIP1 Inhibitor I, RIPK 1 Inhibitor I, Receptor-Interacting Protei, Necrosis Inhibitor II, 5-((1H-Indol-3-yl)methyl)-3-methyl-2-thioxoimidazolidin-4-one; 5-(Indol-3-ylmethyl)-(2-thio-3-methyl)hydantoin, MTH-Trp, Methyl-thiohydantoin-tryptophan, Nec-1, Nec1, Necrosome Inhibitor I, RIP1 Inhibitor I, RIPK 1 Inhibitor I, Receptor-Interacting Protein 1 Inhibitor I

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H13N3OS
Molecular Weight:
259.33
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥99% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
desiccated (hygroscopic)
protect from light

storage temp.

−70°C

General description

A cell-permeable, potent, and selective blocker of necroptosis (EC50 = 494 nM in FADD-deficient Jurkat cells treated with TNF-α), a nonapoptotic necrotic cell death pathway mediated by death-domain receptors (DRs) that offers neuroprotection in a murine model of ischemic brain injury. Exhibits no effect on DR-induced apoptosis. Also acts as a selective and ATP-competitive inhibitor of RIP1 kinase with negligible effect on RIP2 kinase activity. Nec-1 target appears to be a critical common necroptotic step upstream of execution events and downstream of DRs. Inactive control, Nec-1i, is also available (>Cat. No. 480066).

Biochem/physiol Actions

Cell permeable: yes
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Physical form

A 25 mM (10 mg/1.54 ml) sterile-filtered solution of Necrostatin-1 (Cat. No. 480065) in DMSO.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C). Aliquots are stable for up to 1 year at -70°C.

Other Notes

Degterev, A., et al. 2013. Nat. Chem. Biol.9, 192.
Degterev, A., et al. 2012. Cell Death Differ.20, 366.
Christofferson, D.E., et al. 2012. Cell Death Dis.3, e320.
Takahashi, N., et al. 2012. Cell Death Dis.3, e437.
Degterev, A., et al. 2008. Nat. Chem. Biol.4, 313.
Degterev, A., et al. 2005. Nat. Chem. Biol.1, 112.
Muller, A.J., et al. 2005. Nat. Med.11, 312.
Teng, X., et al. 2005. Bioorg. Med. Chem. Lett.15, 5039.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F - (refers to pure substance)

Flash Point(C)

87 °C - (refers to pure substance)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service