Skip to Content
Merck
All Photos(1)

Key Documents

5.04906

Sigma-Aldrich

Cuspin-1

Synonym(s):

Cuspin-1, (5-Bromo-3-pyridinyl)(4-methylphenyl)methanone, Survival Motor Neuron protein Upregulator, Cuspin-1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H10BrNO
CAS Number:
Molecular Weight:
276.13
UNSPSC Code:
12352200

Assay

≥94% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

brown

solubility

DMSO: 10 mg/mL

storage temp.

2-8°C

General description

A cell-permeable pyridinylphenyl-methanone compound that is reported to effectively upregulate SMN (survival of motor neuron) protein level in 3 out of 4 SMA (spinal muscular atrophy) patient-derived fibroblast lines (by 71% to 93% increase in 48 h; 5 µg/mL), but not HT1080, 293T, A549, two human ESC-MN lines, MEF, mESC-MN, rat PC12, or rat ST14A cells. Shown to preferentially stimulate cellular Erk (Thr202/Tyr204) over Akt (Ser473) phosphorylation in SMA fibroblast line 9677 in a time-dependent manner (tmax = 24 h; 10 µg/mL), indicative of Ras-Raf-MEK signaling pathway activation.

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Ras
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Letso, R.R., et al. 2013. ACS Chem. Biol.8, 914.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service