5.04361
INCB 3284 Dimesylate
Synonym(s):
INCB 3284 Dimesylate, N-[2-[[(3R)-1-[trans-4-Hydroxy-4-(6-methoxy-3-pyridinyl)cyclohexyl]-3-pyrrolidinyl]amino]-2-oxoethyl]-3-(trifluoromethyl)-benzamide dimethanesulfonate, INCB 3284, INCB3284
About This Item
Assay
≥98% (TLC)
Quality Level
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
color
off-white
solubility
DMSO: 12 mg/mL
storage temp.
−20°C
InChI
1S/C26H31F3N4O4.2CH4O3S/c1-37-23-6-5-19(14-30-23)25(36)10-7-21(8-11-25)33-12-9-20(16-33)32-22(34)15-31-24(35)17-3-2-4-18(13-17)26(27,28)29;2*1-5(2,3)4/h2-6,13-14,20-21,36H,7-12,15-16H2,1H3,(H,31,35)(H,32,34);2*1H3,(H,2,3,4)/t20-,21?,25?;;/m1../s1
InChI key
PNPNUHKICDECDH-SEBNIYPMSA-N
General description
Biochem/physiol Actions
human CCR2 (hCCR2)
Warning
Reconstitution
Other Notes
Xue, C., et al. 2011. ACS Med. Chem. Lett.2, 450.
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service