Skip to Content
Merck
All Photos(1)

Key Documents

481974

Sigma-Aldrich

NADPH-P450 Oxidoreductase, Human, Recombinant, S. frugiperda

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
UNSPSC Code:
12352202

Assay

≥90% (SDS-PAGE)

Quality Level

form

liquid

specific activity

≥20 units/mg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

solubility

aqueous buffer: soluble

shipped in

wet ice

storage temp.

−70°C

General description

Recombinant, human NADPH-P450 oxidoreductase expressed in S. frugiperda insect cells. A membrane bound flavoprotein containing one mole each of FAD and FMN. Transfers electrons from NADPH to the various isoforms of cytochrome P450. Typically used at ratio 1:10 to 1:2 (reductase:P450) for in vitro reconstitution assays. Suitable for in vitro drug metabolism and toxicology research including reconstituted P450-dependent biotransformation reactions.
Recombinant, human NADPH-P450 oxidoreductase expressed in S. frugiperda insect cells. A membrane-bound flavoprotein containing one mole each of FAD and FMN. Transfers electrons from NADPH to the various isoforms of cytochrome P450. Typically used at ratios ranging from 1:10 to 1:2 (reductase:P450) for in vitro reconstitution assays. Suitable for in vitro drug metabolism and toxicology research including reconstituted P450-dependent biotransformation reactions.

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that will oxidize 1 µmol of cytochrome c per min at 20°C, pH 7.7.

Physical form

In 10 mM potassium phosphate buffer, 1 mM DTT, 100 µM EDTA, 20% glycerol, pH 7.7.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Shen, A.L., et al. 1989. J. Biol. Chem. 264, 7584.
Yamano, S., et al. 1989. Mol. Pharmacol.36, 83.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Donghua Du et al.
Cell stress & chaperones, 26(2), 387-401 (2021-01-07)
Liver damage is the most severe complication of heat stress (HS). Hydrolyzed camel whey protein (CWP) possesses bioactive peptides with obviously antioxidant and anti-inflammatory activities. The current study aims to investigate whether CWP that is hydrolyzed by a simulated gastrointestinal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service