Skip to Content
Merck
All Photos(1)

Key Documents

445821

Sigma-Aldrich

Meriolin 3

A cell-permeable, ATP site-targeting azaindolopyrimidine compound that is reported to inhibit kinase activity.

Synonym(s):

Meriolin 3, 3-((2-Amino)pyrimidin-4-yl)-4-methoxy-1H-pyrrolo[2,3-b]pyridine, Cdk7 Inhibitor VI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H11N5O
CAS Number:
Molecular Weight:
241.25
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C12H11N5O/c1-18-9-3-5-14-11-10(9)7(6-16-11)8-2-4-15-12(13)17-8/h2-6H,1H3,(H,14,16)(H2,13,15,17)

InChI key

GGEWEQJWGGJUHO-UHFFFAOYSA-N

General description

A cell-permeable, ATP site-targeting azaindolopyrimidine compound that is reported to inhibit the kinase activity of CDK9/cyclin T, CDK2/cyclin A, CDK1/cyclin B, CDK5/p25 (IC50 = 6, 11, 170, 170 nM, respectively), DYRK1A, CKIδ/ε, GSK-3α/β (IC50 = 29, 200, and 230 nM, respectively), as well as CDK3/cyclin E, CDK4/cyclin D1, CDK6/cyclin D1, CDK7/cyclin H, and FLT3 (IC50<400 nM), while showing much reduced activity agasint 21 other kinases (IC50 >1 µM). Shown to effectively inhibit cellular CDK substrates phosphorylation and induce apoptosis in SH-SY5Y neuroblastoma cultures (Effective conc. 0.3 to 1 µM) in vitro and retard A4573- and LS174T-derived tumor growths in mice in vivo (1 to 5 mg/kg/day, i.p.). Cell viability tests reveal that Meriolin 3 selectively affects the viability of a panel of 8 cancer lines (IC50 ≤380 nM) in comparison to normal human foreskin fibroblasts (IC50 = 8 µM).

Packaging

Packaged under inert gas

Warning

Toxicity: Regulatory Review (Z)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Echalier, A., et al. 2008. J. Med. Chem.51, 737.
Bettayeb, K., et al. 2007. Cancer Res.67, 8325.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service