Skip to Content
Merck
All Photos(1)

Documents

422000

Sigma-Aldrich

Kenpaullone

A potent, cell-permeable, and reversible inhibitor of glycogen synthase kinase-3β (IC₅₀ = 230 nM), Lck (IC₅₀ = 470 nM), and cyclin-dependent kinases (Cdks).

Synonym(s):

Kenpaullone, 9-Bromo-7,12-dihydroindolo[3,2-d][1]benzazepin-6(5H)-one, NSC-664704

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H11BrN2O
CAS Number:
Molecular Weight:
327.18
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow-brown

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

−20°C

InChI

1S/C16H11BrN2O/c17-9-5-6-14-11(7-9)12-8-15(20)18-13-4-2-1-3-10(13)16(12)19-14/h1-7,19H,8H2,(H,18,20)

InChI key

QQUXFYAWXPMDOE-UHFFFAOYSA-N

General description

A potent cell-permeable and reversible inhibitor of glycogen synthase kinase-3β (IC50 = 230 nM), Lck (IC50 = 470 nM), and cyclin-dependent kinases (Cdks). Inhibits Cdk1/cyclin B (IC50 = 400 nM), Cdk2/cyclin A (IC50 = 680 nM), Cdk2/cyclin E (IC50 = 7.5 µM), and Cdk5/p25 (IC50 = 850 nM). Also inhibits other kinases such as c-Src (IC50 = 15 µM), casein kinase II (IC50 = 20 µM), ERK1 (IC50 = 20 µM), and ERK2 (IC50 = 9 µM). Inhibition is competitive with respect to ATP binding.
A potent, cell-permeable, and reversible inhibitor of glycogen synthase kinase-3β (IC50 = 230 nM), Lck (IC50 = 470 nM), and cyclin-dependent kinases (Cdks). Inhibits Cdk1/cyclin B (IC50 = 400 nM), Cdk2/cyclin A (IC50 = 680 nM), Cdk2/cyclin E (IC50 = 7.5 µM), and Cdk5/p25 (IC50 = 850 nM). Also inhibits other kinases such as c-Src (IC50 = 15 µM), casein kinase II (IC50 = 20 µM), ERK1 (IC50 = 20 µM), and ERK2 (IC50 = 9 µM). Inhibition is competitive with respect to ATP binding.

Biochem/physiol Actions

Cell permeable: yes
Product competes with ATP.
Reversible: yes
Target IC50: 230 nM, 470 nM, 400 nM, 680 nM, 7.5 µM, 850 nM, 15 µM, 20 µM, 20 µM, 9 µM against glycogen synthase kinase-3β, Lck IC50, Cdk1/cyclin B, Cdk2/cyclin A, Cdk2/cyclin E, and Cdk5/p25, c-Src, casein kinase II, ERK1, and ERK2

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Bain, J., et al. 2003. Biochem. J. 371, 199.
Buolamwini, J.K. 2000. Curr. Pharm. Des.6, 379.
Schultz, C., et al. 1999. J. Med. Chem. 42, 2909.
Zaharevitz, D.W., et al. 1999. Cancer Res. 59, 2566.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bryan A Johnson et al.
PLoS pathogens, 18(6), e1010627-e1010627 (2022-06-22)
While SARS-CoV-2 continues to adapt for human infection and transmission, genetic variation outside of the spike gene remains largely unexplored. This study investigates a highly variable region at residues 203-205 in the SARS-CoV-2 nucleocapsid protein. Recreating a mutation found in
Jonathan Bayerl et al.
Cell stem cell, 28(9), 1549-1565 (2021-04-30)
Isolating human MEK/ERK signaling-independent pluripotent stem cells (PSCs) with naive pluripotency characteristics while maintaining differentiation competence and (epi)genetic integrity remains challenging. Here, we engineer reporter systems that allow the screening for defined conditions that induce molecular and functional features of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service