Skip to Content
Merck
All Photos(1)

Key Documents

373273-M

Millipore

Hh Signaling Antagonist XIV, SANT-2

The Hh Signaling Antagonist XIV, SANT-2, also referenced under CAS 329196-48-7, controls the biological activity of Hh pathway. This small molecule/inhibitor is primarily used for Cell Signaling applications.

Synonym(s):

Hh Signaling Antagonist XIV, SANT-2, N-[3-(1H-Benzimidazol-2-yl)-4-chlorophenyl]-3,4,5-tris(ethyloxy)benzamide, Smo Antagonist V

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H26ClN3O4
CAS Number:
Molecular Weight:
479.96
UNSPSC Code:
12352200

Pricing and availability is not currently available.

Assay

≥95% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 100 mg/mL
ethanol: 25 mg/mL

storage temp.

2-8°C

InChI

1S/C26H26ClN3O4/c1-4-32-22-13-16(14-23(33-5-2)24(22)34-6-3)26(31)28-17-11-12-19(27)18(15-17)25-29-20-9-7-8-10-21(20)30-25/h7-15H,4-6H2,1-3H3,(H,28,31)(H,29,30)

InChI key

VQOJFGFKIVFMDH-UHFFFAOYSA-N

General description

A nanomolar Smoothened (smo) antagonist that exhibits 2.3- and 6-fold weaker biological activity than SANT-1 (Cat. No. 559303), respectively, in antagonizing ShhNp- and SAG-stimulated hedgehog pathway activation in HEK 293-based (Shh-LIGHT2) reporter assays. Competitive binding studies show full replacement of Smo-bound cyclopamine (Cat. No. 239803) by SANT-1 and SANT-2 (Ki = 2.4 and 8.4 nM, respectively), while SANT-1 and SANT-2 are shown to compete against Smo-bound SAG-1.3 (Cat. Nos. 566660 & 566661) in an allosteric manner and that only SANT-2, but not SANT-1, is able to completely compete off bound SAG-1.3 (100% vs. 40% replacement by the respective compound at 10 µM).
A nanomolar Smoothened (smo) antagonist that exhibits 2.3- and 6-fold weaker biological activity than SANT-1 (Cat. No. 559303), respectively, in antagonizing ShhNp- and SAG-stimulated hedgehog pathway activation in HEK 293-based (Shh-LIGHT2) reporter assays. Competitive binding studies show full replacement of Smo-bound cyclopamine (Cat. No. 239803) by SANT-1 and SANT-2 (Ki = 2.4 and 8.4 nM, respectively), while SANT-1 and SANT-2 are shown to compete against Smo-bound SAG-1.3 (Cat. Nos. 566660 & 566661) in an allosteric manner and that only SANT-2, but not SANT-1, is able to completely compete off bound SAG-1.3 (100% vs. 40% replacement by the respective compound at 10 µM).

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Rominger, C.M., et al. 2009. J. Pharmacol. Exp. Ther.329, 995.
Rohatgi, R., et al. 2009. Proc. Natl. Acad. Sci. USA106, 3196.
Chen, J.K., et al. 2002. Proc. Natl. Acad. Sci. USA99, 14071.
Frank-Kamenetsky, M., et al. 2002. J. Biol.1, 10.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service