Skip to Content
Merck
All Photos(1)

Key Documents

3660-OP

Sigma-Aldrich

DEPC

OmniPur® Grade, >= 97.5%, for the preparation of RNase-free solutions

Synonym(s):

OmniPur® DEPC, Diethyl Dicarbonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O(COOCH2CH3)2
CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.25

Quality Level

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

clear light yellow

shipped in

wet ice

storage temp.

10-30°C

InChI

1S/C6H10O5/c1-3-9-5(7)11-6(8)10-4-2/h3-4H2,1-2H3

InChI key

FFYPMLJYZAEMQB-UHFFFAOYSA-N

General description

For the preparation of RNase-free solutions.Inhibitor of RNase to protect sensitive RNA molecules. Molecular biology grade.

Specifications:
Assay (with acid): ≥97.5%





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

156.2 °F - closed cup

Flash Point(C)

69 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service