Skip to Content
Merck
All Photos(1)

Documents

345878

Sigma-Aldrich

GGTI-286

A potent, cell-permeable, and selective inhibitor of GGTase I.

Synonym(s):

GGTI-286, N-4-[2(R)-Amino-3-mercaptopropyl]amino-2-phenylbenzoyl-(L)-leucine methyl ester, TFA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H31N3O3S
CAS Number:
Molecular Weight:
429.58
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥90% (HPLC)

form

solid (Glassy)

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A potent, cell-permeable, and selective GGTase-I inhibitor. This methyl ester derivative of GGTI-287 (Cat. No. 345880) is 25-fold more potent (IC50 = 2 µM versus 50 µM) than FTI-277 (Cat. No. 344555) inhibiting the processing of the geranylgeranylated protein Rap1A. Selectively antagonizes oncogenic K-Ras4B (IC50 = 2 µM). GGTI-286 has also been shown to have a significant antiproliferative effect in human malignant glioma cells.
A potent, cell-permeable, and selective inhibitor of GGTase I. This methyl ester derivative of GGTI-287 (Cat. No. 345880) is about 25-fold more potent (IC50 = 2 µM vs. 50 µM) than FTI-277 (Cat. No. 344555) in inhibiting the processing of the geranylgeranylated protein Rap1A. Selectively antagonizes oncogenic K-Ras4B (IC50 = 2 µM). GGTI-286 has also been shown to have a significant antiproliferative effect in human malignant glioma cells.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Geranylgeranyltransferase (GGTase)
Product does not compete with ATP.
Reversible: no
Target IC50: 2 µM against GGTase

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution aliquot and freeze (-70°C). Stock solutions are stable for up to 1 week at -70°C.

Other Notes

Bredel, M., et al. 1998. Neurosurgery 43, 124.
Qian, Y., et al. 1998. Bioorg. Med. Chem.6, 293.
Lerner, E.C., et al. 1997. Oncogene 15, 1283.
Lerner, E.C., et al. 1995. J. Biol. Chem. 270, 26770.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service