Skip to Content
Merck
All Photos(1)

Documents

345872

Sigma-Aldrich

Glial Derived Neurotrophic Factor, Human, Recombinant, E. coli

Synonym(s):

rhGDNF

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

Assay

≥97% (SDS-PAGE)

Quality Level

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

solubility

aqueous buffer: soluble (most)
water: soluble

shipped in

ambient

storage temp.

−20°C

General description

Recombinant, human glial derived neutotrophic factor expressed in E. coli. A heterogeneously glycosylated growth promoting factor that enhances neuronal survival of dopaminergic midbrain neurons. Activates receptor tyrosine kinase Ret and promotes kidney morphogenesis. Also an activator of MAP kinase. Its biological activity is destroyed by reduction of disulfide bonds.
Recombinant, human glial derived neutotrophic factor expressed in E. coli. Growth promoting factor that enhances neuronal survival of dopaminergic midbrain neurons. A heterogeneously glycosylated basic protein. Its biological activity is destroyed by the reduction of disulfide bonds.

Biochem/physiol Actions

fully biologically active when compared to native GDNF by dose-dependent ³H-dopamine uptake assay

Warning

Toxicity: Standard Handling (A)

Preparation Note

The recommended stock solution is 1 µg/100µl. For more dilute samples, add a stabilizing protein such as 0.1% BSA.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Avoid freeze/thaw cycles of solutions. Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Vega, Q.C., et al. 1996. Proc. Natl. Acad. Sci. USA 93, 10657.
Worby, C.A., et al. 1996. J. Biol. Chem. 271, 23619.
Lin, L.F., et al. 1994. J. Neurochem. 63, 758.
Lev-Fen, H., et al. 1993. Science260, 1130.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service