2400
Stat-Matic I Plate Washer, bottle-top dispenser
The StatMatic I automated plate washer provides a convenient & affordable alternative for diagnostic assays & cell culture work.
Synonym(s):
plate washer
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41104923
eCl@ss:
36102014
Recommended Products
manufacturer/tradename
Chemicon®
shipped in
ambient
General description
The StatMatic I provides a convenient and affordable alternative for diagnostic assays and cell culture work. Used with an eight-port disposable manifold, the StatMatic I provides continuous operation not possible with multi-channel pipettes. The eight-port manifold, designed for 96-well microplates, delivers fluids in volumes of 100, 200, 250, or 300μL with well-to-well accuracy of 5% or better.
Constructed of polypropylene, PTFE, glass, and ceramics, the StatMatic I has no exposed glass that can break and requires minimal maintenance. The StatMatic I is compatible with most reagents and chemicals and is designed so that even protein-based reagents won′t cause sticking. The filtered air vent keeps liquids free of contaminants. In addition, the priming line allows recirculation into the dispenser port without removing the bottle.
Constructed of polypropylene, PTFE, glass, and ceramics, the StatMatic I has no exposed glass that can break and requires minimal maintenance. The StatMatic I is compatible with most reagents and chemicals and is designed so that even protein-based reagents won′t cause sticking. The filtered air vent keeps liquids free of contaminants. In addition, the priming line allows recirculation into the dispenser port without removing the bottle.
Legal Information
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
General Lab Use
Signal Word
Danger
Hazard Statements
Precautionary Statements
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service