A cell-permeable pyrimido-isoquinolinone compound that acts as a potent, reversible, and ATP-competitive inhibitor of DNA-PK (IC₅₀ = 280 nM) with ~19-fold selectivity over mTOR (IC₅₀ = 5.3 µM).
A cell-permeable pyrimido-isoquinolinone compound that acts as a potent, reversible, and ATP-competitive inhibitor of DNA-PK (IC50 = 280 nM) with ~19-fold selectivity over mTOR (IC50 = 5.3 µM). Exhibits much reduced or no activity against 43 other commonly studied kinases, including PI 3-K, ATM, and ATR. Shown to induce apoptosis and inhibit mTOR-dependent growth in TSC1-/- murine embryo fibroblasts.
Packaging
Packaged under inert gas
Warning
Toxicity: Standard Handling (A)
Reconstitution
Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.
Other Notes
Ballou, L.M., et al. 2007. J. Biol. Chem.282, 24463. Griffin, R.J., et al. 2005. J. Med. Chem.48, 569.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.