Skip to Content
Merck
All Photos(1)

Key Documents

1.93715

Supelco

Cadmium chloride

anhydrous, for analysis EMPLURA®

Synonym(s):

Cadmium chloride, Cadmium dichloride, Dichlorocadmium

Sign Into View Organizational & Contract Pricing

Select a Size

1937150121
₹1,389.99

₹1,389.99


Estimated to ship on31 March 2025Details


Request a Bulk Order

Select a Size

Change View
1937150121
₹1,389.99

About This Item

Empirical Formula (Hill Notation):
CdCl2
Molecular Weight:
183.32
MDL number:
UNSPSC Code:
12352302

₹1,389.99


Estimated to ship on31 March 2025Details


Request a Bulk Order

grade

anhydrous

vapor density

6.3 (vs air)

vapor pressure

10 mmHg ( 656 °C)

mp

568 °C (lit.)

storage temp.

room temp

SMILES string

Cl[Cd]Cl

InChI

1S/Cd.2ClH/h;2*1H/q+2;;/p-2

InChI key

YKYOUMDCQGMQQO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service