Skip to Content
Merck
All Photos(1)

Key Documents

1.93669

Supelco

Lead(II) nitrate

EMPLURA®

Synonym(s):

Lead(II) nitrate

Sign Into View Organizational & Contract Pricing

Select a Size

1936690521
₹375.00

₹375.00


Estimated to ship on31 March 2025Details


Request a Bulk Order

Select a Size

Change View
1936690521
₹375.00

About This Item

Linear Formula:
Pb(NO3)2
CAS Number:
Molecular Weight:
331.21
MDL number:
UNSPSC Code:
12352302

₹375.00


Estimated to ship on31 March 2025Details


Request a Bulk Order

product line

EMPLURA®

storage temp.

room temp

SMILES string

[Pb](O[N+](=O)[O-])O[N+](=O)[O-]

InChI

1S/2NO3.Pb/c2*2-1(3)4;/q2*-1;+2

InChI key

RLJMLMKIBZAXJO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Repr. 1A - Skin Sens. 1B - STOT RE 1

Target Organs

Blood,Central nervous system,Immune system,Kidney

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service