Skip to Content
Merck
All Photos(1)

Key Documents

1690-OP

Millipore

Acrylamide/Bis-acrylamide

OmniPur® Grade, 40% solution, Suitable for nucleic acid and protein separation sequencing.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105319

Quality Level

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

clear colorless

shipped in

wet ice

storage temp.

2-8°C

General description

Suitable for nucleic acid and protein separation sequencing. Premixed solutions offer standard ratios of acrylamide to bis-acrylamide for convenience and safety. The hazard of handling powdered acrylamide is eliminated with the 40% solution in water. Reconstitute with deionized H2O to desired final percentage. Each pouch makes 1L of 10X TBE Buffer (1X = 0.089M TRIS, 0.089M Borate, 0.002M EDTA). Molecular biology grade.

Specifications:
DNAse: None detected
RNAse: None detected
Protease: None detected





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Warning

Toxicity: Toxic (F)

Other Notes

Specifications
Assay (iodometric)91% min
ColorWhite
FormPowder
Identity (IR-spectrum)Conforms
Water (H2O)1% max
Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Muta. 1B - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Oral

Target Organs

Peripheral nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service