Skip to Content
Merck
All Photos(2)

Key Documents

1.30190

Millipore

HY-LiTE® 2/2A Cuvette Holder

Replacement cuvette holder for HY-LiTE® 2/2A luminometers

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122409
NACRES:
NA.74
Pricing and availability is not currently available.

compatibility

(Compatible with HY-LiTE® 2/2A instrument versions)

storage temp.

2-30°C

General description

Replacement or spare cuvette holder for HY-LiTE® 2/2A instruments. The cuvette is used to hold the activated pen during testing.

Application

For use as a spare part or replacement of the cuvette holder originally included with the luminometer.

Legal Information

HY-LITE is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service