1.18461
Nickel Test Refill pack
for use with MCOLORTEST®
Synonym(s):
Nickel Visual Test Kit Refills
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
feature
refill
Quality Level
specific analyte(s)
nickel
compatibility
for use with MCOLORTEST®
detection method
colorimetric
storage temp.
15-25°C
General description
When the reagents of a MQuant® test (formally MColortest®) are used up, the kit can be replenished by ordering the appropriate refill pack. Refill packs are available for a range of test kits, making it a particularly economical and ecological system.
This refill pack contains all reagents for the MQuant® test kits 1.14783 and 1.14420 for the colometric evaluation of nickel.
This refill pack contains all reagents for the MQuant® test kits 1.14783 and 1.14420 for the colometric evaluation of nickel.
for 114783, 114420 MColortest®
Legal Information
MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1 - STOT RE 2 Oral - STOT SE 3
Target Organs
Respiratory system, Thyroid
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
48.2 °F
Flash Point(C)
9 °C
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service