Key Documents
1.18330
Supelco
Turbidimeter
About This Item
UNSPSC Code:
47101516
General description
The Turbiquant 1500 T is suitable for all routine measurements in the laboratory. The instrument automatically selects the correct measuring range. The optional continuous flow cell makes it possible to run pressureless continuous-flow measurements. The instrument contains an IR-LED (infrared) light source with a wavelength of 860 nm for standard-compliant measurements according to EN ISO 7027.
incl. universal power supply/plug, 3 empty cells, manual, handy hints 1500 IR 0 - 1000 NTU Turbiquant
Legal Information
TURBIQUANT is a trademark of Merck KGaA, Darmstadt, Germany