116815-M
Adenosine 3′,5′-cyclic Monophosphorothioate, Sp-Isomer, Triethylammonium Salt
A cell-permeable cAMP analog that binds cAMP-receptor proteins.
Synonym(s):
Adenosine 3′,5′-cyclic Monophosphorothioate, Sp-Isomer, Triethylammonium Salt, Adenosine 3ʹ,5ʹ-cyclic Phosphorothioate-Sp, Sp-cAMPS, TEA
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Assay
≥98% (HPLC)
form
crystalline solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
desiccated (hygroscopic)
solubility
water: 20 mg/mL
storage temp.
−20°C
General description
A cell-permeable cAMP analog that binds cAMP-receptor proteins. Induces the guanine-nucleotide exchange factor activity of GEFI (also known as Epac) and GEFII towards Raf1, both in vivo and in vitro. Known to cause a decrease in cytosolic pH. Note: 10 µmol = 4.47 mg.
A cell-permeable cAMP analog that binds cAMP-receptor proteins. Induces the guanine-nucleotide-exchange factor activity of GEFI, also known as Epac (exchange protein directly activated by cAMP), and GEFII towards Rap1 both in vivo and in vitro. Known to cause a decrease in cytosolic pH. Note: 10 µmol = 4.47 mg. λmax = 258 nm; εmax = 15,000 M-1cm-1 (pH 7..0).
Note: 10 µmol = 4.47 mg.
Biochem/physiol Actions
Primary Target
cAMP-receptor proteins
cAMP-receptor proteins
Warning
Toxicity: Standard Handling (A)
Reconstitution
Following reconstitution, aliquot and freeze (-20°C). Aqueous stock solutions are stable for up to 1 month at -20°C.
Other Notes
Kawasaki, H., et al. 1998. Science 282, 2275.
de Rooij, J., et al. 1998. Nature 396, 474.
Kaji, H., et al. 1992. Biochem. Biophys. Res. Commun.182, 1356.
Sugimoto, T., et al. 1992. J. Cell Physiol. 152, 28.
Van Der Kloot, W., and Branisteanu, D.D. 1992. Pflugers Arch.420, 336.
Takuma, T., and Ichida, T. 1991. J. Biochem.110, 292.
Dostmann, W.R., et al. 1990. J. Biol. Chem.265, 10484.
de Rooij, J., et al. 1998. Nature 396, 474.
Kaji, H., et al. 1992. Biochem. Biophys. Res. Commun.182, 1356.
Sugimoto, T., et al. 1992. J. Cell Physiol. 152, 28.
Van Der Kloot, W., and Branisteanu, D.D. 1992. Pflugers Arch.420, 336.
Takuma, T., and Ichida, T. 1991. J. Biochem.110, 292.
Dostmann, W.R., et al. 1990. J. Biol. Chem.265, 10484.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service