Skip to Content
Merck
All Photos(1)

Documents

1.08048

Supelco

Carbonate Hardness Test

for use with MQUANT®

Synonym(s):

Carbonate Hardness Visual Test Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

specific analyte(s)

carbonate, hydrogen carbonate

Quality Level

compatibility

for use with MQUANT®

detection method

titrimetric

storage temp.

15-25°C

General description

The MQuant® Carbonate Hardness test (formally MColortest®) uses a graduated syringe to determine carbonate hardness as a measure of the buffering capacity of water which can also be expressed as acid-binding capacity or total alkalinity.

Method
Hydrogen carbonate and carbonate ions are titrated with hydrochloric acid against a mixed indicator. At the titration end-point (at pH 4.3) the color changes to red. The carbonate hardness (acid capacity) is determined from the consumption of titration solution.

This test can be used in
  • Surface water, seawater
  • Drinking water and mineral water (after degassing)
  • Aquarium water, waters from aquaculture
  • Industrial water, process water
  • Boiler and boiler feed water, cooling water

Analysis Note

Application test: conforms

Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameCorrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Met. Corr. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service