Skip to Content
Merck
All Photos(1)

Key Documents

1.00787

Supelco

Positioning Aid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115311

technique(s)

photometry: suitable

compatibility

for use with Spectroquant®

storage temp.

no temp limit

General description

for 10 mm plastic cuvette Spectroquant®

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service