Skip to Content
Merck
All Photos(2)

Documents

06-942-AF647

Sigma-Aldrich

Anti-acetyl-Histone H3 (Lys9) Antibody, Alexa Fluor 647 Conjugate

from rabbit, ALEXA FLUOR 647

Synonym(s):

Lys9-acetylated

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

ALEXA FLUOR 647

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, mouse, human

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

acetylation (Lys9)

Gene Information

human ... HIST1H3F(8968)

Related Categories

General description

Histone H3 is one of the five main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N-terminal tail, H3 is involved with the structure of the nucleosomes of the ′beads on a string′ structure. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of epigenetic modifications that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine. Histone H3 variants (H3.1, H3.2 and H3.3) have been implicated in the epigenetic memory of cellular state. Genome-wide patterns of H3 are dependent on amino acid sequence and change with cellular differentiation at developmentally regulated loci.

Specificity

Histone H3 acetylated at position 9.

Immunogen

KLH-conjugated synthetic peptide (ARTKQTARK(Ac)STG-C) corresponding to amino acids 1-12 of Histone H3 with acetylated Lys9.

Application

Anti-acetyl-Histone H3 (Lys9), Alexa Fluor 647 Conjugate Antibody, Cat. No. 06-942-AF647, is a highly specific rabbit polyclonal antibody that targets histone H3 Lys9 acetylation (H3K9ac) and has been tested in Flow Cytometry and Immunocytochemistry.
Flow Cytometry Analysis: 2 µL from a representative lot detected cell population with histone H3 Lys9 acetylation (H3K9ac) among one million HEK293 cells.

The unconjugated antibody (Cat. No. 06-492) is shown to be suitable also for Western blotting, dot blot, flow cytometry, chromatin immunoprecipitation (ChIP) and ChIP-seq applications.
Research Category
Epigenetics & Nuclear Function

Quality

Evaluated by Immunocytochemistry in HeLa cells.

Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected histone H3 Lys9 acetylation (H3K9ac) in HeLa cells.

Target description

~17 kDa observed.

Physical form

Protein A purified.
Purified rabbit polyclonal antibody in PBS with 1.5% BSA with 0.05% sodium azide

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service