Skip to Content
Merck
All Photos(3)

Documents

05-1469

Sigma-Aldrich

Anti-NFĸB Antibody, p65, clone 1G10.2

clone 1G10.2, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

1G10.2, monoclonal

species reactivity

bovine, human, mouse, rat

technique(s)

immunocytochemistry: suitable
immunofluorescence: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... RELA(5970)

General description

NFĸB (Nuclear Factor kappa B) represents a group of transcription factors that are structurally related and evolutionarily conserved proteins related to the proto-oncogene c-Rel. The five members in mammals includes Rel (cRel), RelA (p65), RelB, NFĸB1 (p50 and its precursor p105), and NFĸB2 (p52 and its precursor p100)1, 2. NFĸB/Rel proteins exist as homo- or heterodimer complexes.
In a majority of unstimulated cells, the NFĸB transcription factors are bound to the inhibitory IĸB proteins and retained in the cytoplasm in their inactive form. Upon stimulation by multiple inducers, IĸBα is rapidly phosphorylated and degraded. This results in the release of the NFĸB complex, most commonly the p105/p65 heterodimer. The p105 subunit is cleaved into its active p50 form. This cleavage exposes the NLS sequence on the p50 subunit. The p50/p65 heteroduplex then translocates to the nucleus where it activates gene transcription.

Specificity

Detects NFĸB p65.
Expected to cross-react with bovine based on complete sequence homology.
Other species have not been tested.

Immunogen

Epitope: NLS
Synthetic peptide corresponding to the NLS region of human NFĸB p65.

Application

Anti-NFĸB Antibody, p65, clone 1G10.2 detects level of NFĸB & has been published & validated for use in WB, IF & IC.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Quality

Routinely evaluated by Western Blot.

Western Blot Analysis:
This lot detected NFB, p65 at 1:1,000 dilution in Jurkat cell lysate resolved via SDS-PAGE and transferred to PVDF.

Target description

65 kDa

Physical form

Format: Purified
Protein A Purified
Purified mouse monoclonal in 0.1M Tris-Glycine (pH 7.4) with 150mM NaCl and 0.05% NaN3.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.
Handling Recommendations: Upon receipt, and prior to removing the cap, centrifuge the vial and gently mix the solution.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service