Skip to Content
Merck
All Photos(4)

Documents

03-903

Sigma-Aldrich

GFP, FLAG-tagged SNRNP70 mRNA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.52

packaging

1 ea of 50 μg

Quality Level

manufacturer/tradename

Millipore

growth mode

N/A

technique(s)

RIP: suitable
cell culture | mammalian: suitable
flow cytometry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

shipped in

dry ice

storage temp.

-65 to -96°C

Application

Both mRNAs and non-coding RNAs interact with RNA-binding proteins and regulate critical cellular processes and their dysregulation contributes to multiple diseases. Thus, analyzing RNA-protein interactions are critical for understanding the mechanism of diseases and designing clinical strategies.
RNA-binding protein immunoprecipitation (RIP) has been used to interrogate RNA binding proteins (RBP) associated with specific RNA molecules. However, the experiments are not feasible because of the unavailability of antibodies for the particular RBP or low expression of the RBP. To overcome these issues epitope tags have been used for RIP experiments.

Features and Benefits

The GFP, FLAG-tagged SNRNP70 mRNA was developed to enable to perform control RIP experiments with RIPAb+ FLAG (Cat. No. 03-901) or RIPAb+ GFP (Cat. No. 03-902) RIP validated antibody and primer kits, and Magna RIP kit (Cat. # 17-700).

Storage and Stability

Store at -80ºC

Other Notes

The high amount of GFP-FLAG-tagged SNRNP70 protein is expressed immediately after transfection to cells and the well-known specific interaction of SNRNP70 and U1snRNA can be used for control RIP experiments with anti-GFP or anti-FLAG antibodies.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service