Skip to Content
Merck
All Photos(1)

Key Documents

MAN640940179

Manchester Organics

[2,6-Difluoro-4-(hydroxymethyl)phenyl]boronic acid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
187.94
MDL number:

Iupac Name

(2,6-difluoro-4-(hydroxymethyl)phenyl)boronic acid

Inchi Code

1S/C7H7BF2O3/c9-5-1-4(3-11)2-6(10)7(5)8(12)13/h1-2,11-13H,3H2

InChI key

MJXZDKYAMFZLML-UHFFFAOYSA-N

Country of Origin

GB

Product Link

https://www.manchesterorganics.com/N55075

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library