Skip to Content
Merck
All Photos(1)

Key Documents

KEYH2F6215A3

Key Organics / BIONET

ethyl 1-(2,2-difluoroethyl)piperidine-3-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
221.25
MDL number:

Iupac Name

ethyl 1-(2,2-difluoroethyl)-3-piperidinecarboxylate

Inchi Code

1S/C10H17F2NO2/c1-2-15-10(14)8-4-3-5-13(6-8)7-9(11)12/h8-9H,2-7H2,1H3

InChI key

OABUDWSXAXKABV-UHFFFAOYSA-N

Purity

95%

Country of Origin

GB

Physical Form

Liquid

Product Link

https://www.keyorganics.net/sku/NS-00135

Pictograms

Exclamation mark

Signal Word

Warning

MSDS

https://www.keyorganics.net/sds/NS-00135


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library