ENA161209224
Enamine
1-{[1-(difluoromethyl)-1H-imidazol-2-yl]methyl}piperazine trihydrochloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
1-{[1-(difluoromethyl)-1H-imidazol-2-yl]methyl}piperazine trihydrochloride
Inchi Code
1S/C9H14F2N4.3ClH/c10-9(11)15-6-3-13-8(15)7-14-4-1-12-2-5-14;;;/h3,6,9,12H,1-2,4-5,7H2;3*1H
InChI key
XPRWIHNQWRXFPK-UHFFFAOYSA-N
storage temp.
RT
Salt Data
Cl
Purity
95
Country of Origin
UA
Shipping Temperature
Normal
Physical Form
powder
Product Link
https://www.enaminestore.com/catalog/EN300-24506
Signal Word
Warning
Hazard Statements
MSDS
https://www.enaminestore.com/catalog/EN300-24506
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service