COMH4211D3FE
Combi-Blocks, Inc.
Ethyl 5-acetyl-2-ethoxybenzoate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
ethyl 5-acetyl-2-ethoxybenzoate
Inchi Code
1S/C13H16O4/c1-4-16-12-7-6-10(9(3)14)8-11(12)13(15)17-5-2/h6-8H,4-5H2,1-3H3
InChI key
YXLALZDDWWTPFS-UHFFFAOYSA-N
storage temp.
Refrigerated
Purity
98%
Country of Origin
CN
Product Link
https://www.combi-blocks.com/cgi-bin/find.cgi?QF-7852
MSDS
https://www.combi-blocks.com/cgi-bin/find.cgi?QF-7852
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service