COM448684954
Combi-Blocks, Inc.
2-(2,5-Dimethylphenyl)phenol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
2',5'-dimethyl[1,1'-biphenyl]-2-ol
Inchi Code
1S/C14H14O/c1-10-7-8-11(2)13(9-10)12-5-3-4-6-14(12)15/h3-9,15H,1-2H3
InChI key
JYTQBTDWTJXTSS-UHFFFAOYSA-N
Purity
95%
Country of Origin
CN
Product Link
https://www.combi-blocks.com/cgi-bin/find.cgi?YA-1504
MSDS
https://www.combi-blocks.com/cgi-bin/find.cgi?YA-1504
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.