Skip to Content
Merck
All Photos(1)

Key Documents

COM448615590

Combi-Blocks, Inc.

2-Benzyloxy-5-methylpyridine-3-boronic acid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
243.07
MDL number:

Iupac Name

2-(benzyloxy)-5-methyl-3-pyridinylboronic acid

Inchi Code

1S/C13H14BNO3/c1-10-7-12(14(16)17)13(15-8-10)18-9-11-5-3-2-4-6-11/h2-8,16-17H,9H2,1H3

InChI key

NVGSTMYZWMKVPE-UHFFFAOYSA-N

storage temp.

Freezer

Purity

98%

Country of Origin

CN

Product Link

https://www.combi-blocks.com/cgi-bin/find.cgi?BB-6594

MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?BB-6594


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library