C12H316CF35A
Chemcia Scientific, LLC
6-Bromo-8-hydroxy-imidazo[1,2-a]pyrazine-2-carboxylic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
6-bromo-8-oxo-7,8-dihydroimidazo[1,2-a]pyrazine-2-carboxylic acid
Inchi Code
1S/C7H4BrN3O3/c8-4-2-11-1-3(7(13)14)9-5(11)6(12)10-4/h1-2H,(H,10,12)(H,13,14)
InChI key
FQRBTSCZBOGGTE-UHFFFAOYSA-N
storage temp.
0-5
Purity
95%
Country of Origin
US
Shipping Temperature
Ambient
Physical Form
Off-White Solid
Product Link
http://www.chemcia.com/product_fl02.asp?id=8244
MSDS
http://www.chemcia.com/product_fl02.asp?id=8244
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service