Skip to Content
Merck
All Photos(1)

Documents

S-101

Supelco

5-Fluoro PB-22 3-carboxyindole metabolite solution

100 μg/mL in acetonitrile, certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H16FNO2
CAS Number:
Molecular Weight:
249.28
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology
forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

O=C(O)C1=CN(CCCCCF)C2=CC=CC=C21

InChI

1S/C14H16FNO2/c15-8-4-1-5-9-16-10-12(14(17)18)11-6-2-3-7-13(11)16/h2-3,6-7,10H,1,4-5,8-9H2,(H,17,18)

InChI key

DYLIZXRQZRUDLA-UHFFFAOYSA-N

General description

5-Fluoro PB-22 3-carboxyindole is a major metabolite of the synthetic cannabinoid 5-Fluoro PB-22, also known as 5F-PB-22. This certified Snap-N-Spike® solution is suitable as starting material for calibrators or controls in synthetic cannabinoid GC/MS or LC/MS methods for urine drug testing, forensic analysis, or clinical toxicology applications.

For those synthetic cannabinoids currently controlled at the federal level, our certified solution standards of these substances are DEA-exempt, allowing laboratories to place orders without additional regulatory paperwork. For US customers, state specific regulatory requirements may apply.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service