852467C
Avanti
C18(Plasm)-18:1 PC
Avanti Research™ - A Croda Brand
Synonym(s):
1-(1-Enyl-stearoyl)-2-oleoyl-GPC, 1-O-1′-(Z)-octadecenyl-2--(9Z-octadecenoyl)-sn-glycero-3-phosphocholine; GPCho(18:0p/18:1(9Z)); 110629
About This Item
Recommended Products
Assay
>99% (TLC)
form
liquid
packaging
pkg of 1 × 5 mL (852467C-10mg)
pkg of 1 × 5 mL (852467C-25mg)
pkg of 1 × 5 mL (852467C-5mg)
manufacturer/tradename
Avanti Research™ - A Croda Brand
concentration
1 mg/mL (852467C-5mg)
2 mg/mL (852467C-10mg)
5 mg/mL (852467C-25mg)
lipid type
phospholipids
cardiolipins
shipped in
dry ice
storage temp.
−20°C
Application
Packaging
Legal Information
also commonly purchased with this product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3
Target Organs
Liver,Kidney, Respiratory system
Storage Class Code
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
WGK 3
Flash Point(F)
does not flash
Flash Point(C)
does not flash
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service