Skip to Content
Merck
All Photos(2)

Documents

850526P

Avanti

Facade-TFA1

Facade-TFA1, powder

Synonym(s):

Tandem Facial Amphiphile-1; 4-((3R,5R,8R,9S,10S,12S,13R,14S,17R)-3,12-bis(((2R,3R,4R,5S,6R)-3,4-dihydroxy-6-(hydroxymethyl)-5-(((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-yl)oxy)-10,13-dimethylhex

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C101H170N2O46
CAS Number:
Molecular Weight:
2148.42
UNSPSC Code:
12161902
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

mol wt

2148.42 g/mol

packaging

pkg of 1 × 25 mg (850526P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850526P

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC[C@@H]1[C@@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@H](O[C@@H]3CC[C@@]4(C)[C@@H](C3)CC[C@@H]5[C@@H]4C[C@H](O[C@@H]6O[C@H](CO)[C@@H](O[C@H]7O[C@H](CO)[C@@H](O)[C@H](O)[C@H]7O)[C@H](O)[C@H]6O)[C@@]8(C)[C@H]5CC[C@@H]8C(CCC(N(C)CCCN(

General description

Facade-TFA1/tandem facial amphiphiles (TFAs) consists of a pair of maltose-functionalized deoxycholate units. TFAs are long and can be readily produced in large quantities.
Tandem facial amphiphile (TFA) consists of a pair of maltose-functionalized deoxycholate units. It stabilizes membrane proteins.

Packaging

5 mL Amber Glass Screw Cap Vial (850526P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
Facade is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tandem facial amphiphiles for membrane protein stabilization
Chae PS, et al.
Journal of the American Chemical Society, 132(47), 16750-16752 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service