Skip to Content
Merck
All Photos(2)

Documents

840499C

Avanti

16:0-20:4 PG

1-palmitoyl-2-arachidonoyl-sn-glycero-3-phospho-(1′-rac-glycerol) (sodium salt), chloroform

Synonym(s):

1-hexadecanoyl-2--(5Z,8Z,11Z,14Z-eicosatetraenoyl)-sn-glycero-3-phospho-(1′racglycerol) (sodium salt); PAPG; PG(16:0/20:4(5Z,8Z,11Z,14Z)); 110651

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H74O10PNa
CAS Number:
Molecular Weight:
792.99
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (840499C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 840499C

concentration

10 mg/mL (840499C-25mg)

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

InChI

1S/C42H75O10P.Na/c1-3-5-7-9-11-13-15-17-18-19-20-22-24-26-28-30-32-34-42(46)52-40(38-51-53(47,48)50-36-39(44)35-43)37-49-41(45)33-31-29-27-25-23-21-16-14-12-10-8-6-4-2;/h11,13,17-18,20,22,26,28,39-40,43-44H,3-10,12,14-16,19,21,23-25,27,29-38H2,1-2H3,(H,47,48);/q;+1/p-1/b13-11-,18-17-,22-20-,28-26-;/t39?,40-;/m1./s1

InChI key

MRDLRFLJUGDCSO-UQAZZXMVSA-M

General description

16:0-20:4 PG or 1-palmitoyl-2-arachidonoyl-sn-glycero-3-phospho-(1′-rac-glycerol) is a sodium salt of glycerophospholipid, with substitution of palmitic acid, arachidonic acid and phospho-rac-(1-glycerol) at the sn-1, sn-2 and sn-3 positions respectively, in the glycerol backbone.

Application

16:0-20:4 PG or 1-palmitoyl-2-arachidonoyl-sn-glycero-3-phospho-(1′-rac-glycerol) (sodium salt) has been used in liposomes to study its effect on the proliferation of mouse keratinocytes, in vitro.

Packaging

5 mL Clear Glass Sealed Ampule (840499C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ding Xie et al.
PloS one, 9(9), e107119-e107119 (2014-09-19)
We have previously shown that liposomes composed of egg-derived phosphatidylglycerol (PG), with a mixed fatty acid composition (comprising mainly palmitate and oleate), inhibit the proliferation and promote the differentiation of rapidly dividing keratinocytes, and stimulate the growth of slowly proliferating

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service