Skip to Content
Merck
All Photos(1)

Key Documents

810301P

Avanti

TopFluor sphingosine

Avanti Research - A Croda Brand

Synonym(s):

(E)-(2S,3R)-2-Amino-13-( dipyrrometheneboron difluoride)-tridec-4-ene-1,3-diol

Sign Into View Organizational & Contract Pricing

Select a Size

250 μG
₹35,419.40

₹35,419.40


Estimated to ship on25 August 2025Details


Request a Bulk Order

Select a Size

Change View
250 μG
₹35,419.40

About This Item

Empirical Formula (Hill Notation):
C26H40BF2N3O2
CAS Number:
Molecular Weight:
475.42
UNSPSC Code:
12352211
NACRES:
NA.79

₹35,419.40


Estimated to ship on25 August 2025Details


Request a Bulk Order

Assay

99% (TLC)

form

powder

packaging

package of 1 × 250 μg

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

Packaging

5 mL Amber Glass Screw Cap Vial (810301P-250UG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
TopFluor is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service