Skip to Content
Merck
All Photos(1)

Key Documents

ATCH99D3B6BD

Atlantic Research Chemicals Ltd

N,N-Diethyl-P-toluidine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
163.26
MDL number:
Pricing and availability is not currently available.

Iupac Name

N,N-diethyl-4-methylaniline

Inchi Code

1S/C11H17N/c1-4-12(5-2)11-8-6-10(3)7-9-11/h6-9H,4-5H2,1-3H3

InChI key

HKJNHYJTVPWVGV-UHFFFAOYSA-N

Purity

95

Country of Origin

GB

Product Link

https://atlantic-chemicals.com/product/?code=AN053464

MSDS

https://atlantic-chemicals.com/product/?code=AN053464


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library